BR95111
BRAND® special pegs for draining rack
Synonym(s):
Accessories, Draining rack, Special pegs
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
General description
The BRAND® special pegs for the draining rack are crafted from PS and are specifically designed to accommodate narrow-mouthed or small vessels, such as test tubes, small volumetric flasks, and graduated cylinders. These pegs provide a secure and stable hold, ensuring that your vessels drain effectively while minimizing the risk of spills or breakage. Their thoughtful design allows for efficient use of space on the draining rack, making them an essential accessory for laboratory environments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)