Skip to Content
Merck
All Photos(1)

Key Documents

BR708132

Flexible discharge tubing with recirculation valve for Dispensette® S, Dispensette® S Organic, and Dispensette S® Trace Analysis

1,2, 5 and 10 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

PTFE

description

coiled, with safety handle

manufacturer/tradename

BRAND 708132

packaging

1 ea of

L

~800 mm

capacity

1,2, 5 and 10 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

PTFE, coiled, length approx. 800 mm, with safety handle.
Not suitable for HF!

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service