Skip to Content
Merck
All Photos(2)

Key Documents

932426

Sigma-Aldrich

4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine

≥99% (HPLC)

Synonym(s):

4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine, 2T-NATA, 2-TNATA, 2TNATA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H48N4
CAS Number:
Molecular Weight:
897.11
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

grade

sublimed grade

Quality Level

description

μh ≈ 3.0 x 10-6 cm2 V−1 s−1

Assay

≥99% (HPLC)

loss

0.5% TGA, > 350 °C (weight loss)

mp

247 °C

solubility

THF: soluble
chloroform: soluble

fluorescence

λem 490 nm±10 nm in THF

Orbital energy

HOMO 5.1 eV 
LUMO 2.2 eV 

λ

in THF

UV absorption

λ: 326 nm±5 nm Amax

SMILES string

c1ccc(cc1)N(c2ccc(cc2)N(c3ccc(cc3)N(c4ccccc4)c5ccc6ccccc6c5)c7ccc(cc7)N(c8ccccc8)c9ccc%10ccccc%10c9)c%11ccc%12ccccc%12c%11

InChI

1S/C66H48N4/c1-4-22-55(23-5-1)68(64-31-28-49-16-10-13-19-52(49)46-64)61-40-34-58(35-41-61)67(59-36-42-62(43-37-59)69(56-24-6-2-7-25-56)65-32-29-50-17-11-14-20-53(50)47-65)60-38-44-63(45-39-60)70(57-26-8-3-9-27-57)66-33-30-51-18-12-15-21-54(51)48-66/h1-48H

InChI key

KDOQMLIRFUVJNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine can be used in various applications in organic electronics and materials chemistry such as organic Light-Emitting Diodes (OLEDs), organic Photovoltaics(OPVs), organic Field-Effect Transistors (OFETs), dye-sensitized solar Cells(DSSCs), as a hole transport material. It can be utilized in the assembly of supramolecular structures or coordination polymers, offering opportunities for the design and synthesis of functional materials.
Injection Layer (HIL) in Organic Light Emitting Diode (OLED).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and electroluminescent properties of 9,10-diphenylanthracene containing 9H-carbazole derivatives for blue organic light-emitting diodes.
Lee S B, et al.
Synth. Meth. Organomet. Inorg. Chem., 203, 174-179 (2015)
Beomjin Kim et al.
Journal of nanoscience and nanotechnology, 14(8), 6404-6408 (2015-05-06)
New hole injection materials for OLED based on phenothiazine were synthesized, and the electro-optical properties of synthesized materials were examined by through UV-Vis. and photoluminescence spectroscopy as well as cyclic voltammetry. 1-NDD-t-BPBP and 2-NDD-t-BPBP showed Tg of 180 and 177
Synthesis and electroluminescent properties of 9,10-diphenylanthracene containing 9H-carbazole derivatives for blue organic light-emitting diodes.
Lee S B, et al.
Synthetic Metals, 203, 174-179 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service