Skip to Content
Merck
All Photos(1)

Key Documents

922307

Sigma-Aldrich

Poly(lactide-alt-glycolide)

average Mn 10000, lactide:glycolide 50:50

Synonym(s):

PLLA-alt-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H6O4)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

feed ratio

lactide:glycolide 50:50

mol wt

average Mn 10000

color

white to beige

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(lactide-alt-glycolide) contains alternating sequence of lactide and clycolide in the polymer chain, unlike Poly(lactide-co-glycolide) (PLGA). The molar ratio of lactic to glycolic units in the polymer is exactly 1:1. Poly(lactide-alt-glycolide) has improved solutibilty in a broad range of organic solvents such as acetonitrile, acetone, dioxane, DCM and THF, providing great convenience for drug delivery researches and applications. Traditional PLGA polymers have a random distribution of glycolide and lactide units which impacts solutibilty. Compared to random PLGA with LA:GA ratios of 50/50, alternative PLGA undergoes hydrolysis in vivo with minimal changes in pH and a lower amount of acidic by-products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Washington et al.
Acta biomaterialia, 65, 259-271 (2017-11-05)
Controlling the backbone architecture of poly(lactic-co-glycolic acid)s (PLGAs) is demonstrated to have a strong influence on the production and release of acidic degradation by-products in microparticle matrices. Previous efforts for controlling the internal and external accumulation of acidity for PLGA
Ji Hoon Jeong et al.
Journal of biomaterials science. Polymer edition, 14(1), 1-11 (2003-03-15)
A series of amphipathic graft copolymers composed of poly(L-lysine) (PLL) as the cationic polymer backbone and biodegradable poly(D,L-lactic-co-glycolic acid) (PLGA) as the grafting chains were synthesized and characterized. The terminal group of PLGA was activated and chemically conjugated to the
Lin Yu et al.
Biomacromolecules, 12(4), 1290-1297 (2011-03-03)
This paper reports the influence of sequence structures of block copolymers composed of poly(lactic acid-co-glycolic acid) (PLGA) and poly(ethylene glycol) (PEG) on their thermogelling aqueous behaviors. A series of thermogelling PLGA-PEG-PLGA triblock copolymers with similar chemical compositions and block lengths

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service