Skip to Content
Merck
All Photos(1)

Key Documents

901592

Sigma-Aldrich

Sulfur dioxide solution

0.5 M in THF

Synonym(s):

Sulfurous anhydride, Sulfurous oxide, Sulphur dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

0.5 M in THF

density

0.8935 g/mL

SMILES string

[S](=O)=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfur dioxide solution can be used as a reducing agent for the reduction of:
  • Pentavalent arsenic to trivalent arsenic in the presence of sulfuric acid.
  • Graphene oxide to graphene in the presence of water.
Valuable reagent offered as a solution in tetrahydrofuran for more convenient handling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

0.5 °F

Flash Point(C)

-17.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service