Skip to Content
Merck
All Photos(1)

Documents

809594

Sigma-Aldrich

11-Deoxycortisol-2,3,4-13C3 solution

100 μg/mL in methanol, ≥98 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C18H30O4
Molecular Weight:
349.44
EC Number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥97% (CP)

form

liquid

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

C[C@]1(C[13C]2)C(CCC3C1CC[C@@]4(C)C3CC[C@]4(O)C(CO)=O)=[13C][13C]2=O

InChI

1S/C21H27O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h15-17,22,25H,3-4,6-10,12H2,1-2H3/t15?,16?,17?,19-,20-,21-/m0/s1/i5+1,11+1,14+1

InChI key

BTJLPTVOOCUWBT-HSCQNOFGSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service