778451
L-Cysteine hydrochloride monohydrate
Wacker Chemie AG, 98.5-101.0%
Synonym(s):
Cysteine hydrochloride monohydrate, Cysteine monohydrate hydrochloride
About This Item
Recommended Products
Assay
98.5-101.0%
form
powder or crystals
optical activity
[α]/D 5.7 to 6.8°
reaction suitability
reaction type: solution phase peptide synthesis
manufacturer/tradename
Wacker Chemie AG
impurities
≤0.2% any ninhydrin positive substance each (total ≤1.0%)
≤0.5% ninhydrin positive substances: impurity A
≤200 ppm ammonium
ign. residue
≤0.1%
loss
8-12% loss on drying
solubility
water: 100 mg/mL at 25 °C
anion traces
sulfate (SO42-): ≤0.030%
cation traces
Fe: ≤10 ppm
suitability
clear, colorless for appearance of solution
application(s)
peptide synthesis
SMILES string
O.Cl.N[C@@H](CS)C(O)=O
InChI
1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1
InChI key
QIJRTFXNRTXDIP-JIZZDEOASA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Biochem/physiol Actions
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service