Skip to Content
Merck
All Photos(2)

Documents

763357

Sigma-Aldrich

Chloro(1,5-cyclooctadiene)copper(I) dimer

95%

Synonym(s):

[Cu(COD)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24Cl2Cu2
CAS Number:
Molecular Weight:
414.36
EC Number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

core: copper
reagent type: catalyst

mp

120-125 °C

SMILES string

[Cl+]1[Cu-][Cl+][Cu-]1.C2CC=CCCC=C2.C3CC=CCCC=C3

InChI

1S/2C8H12.2Cl.2Cu/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;;;;/q;;2*+1;2*-1/b2*2-1-,8-7-;;;;

InChI key

QKMKFTHUKVKWFX-MIXQCLKLSA-N

General description

Versatile catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyhedron, 10, 2293-2293 (1991)
Chemistry & Industry, 15, 743-743 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service