Skip to Content
Merck
All Photos(2)

Key Documents

74893

Sigma-Aldrich

Octatriacontane

≥95.0% (GC)

Synonym(s):

n-Octatriacontane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)36CH3
CAS Number:
Molecular Weight:
535.03
Beilstein:
1713496
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

flakes

mp

77-79 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C38H78/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-38H2,1-2H3

InChI key

BVKCQBBZBGYNOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Leticia Narciso-Ortiz et al.
Environmental science and pollution research international, 27(18), 23035-23047 (2020-04-26)
Plastic and hydrocarbon pollution in aquatic ecosystems is a worldwide reality and serious concern today. Plastic debris presents a threat to ecosystems and organisms. Hydrocarbons are also considered priority pollutants. The hydrophobicity of the polymer in combination with the high
D Walzthöny et al.
Ultramicroscopy, 6(3), 259-266 (1981-01-01)
Experiments with yeast plasmalemma fracture faces, produced at -196 degrees C and exposed to pure water vapor before replication, showed a "specific decoration" with ice crystals of those pits in the extraplasmatic face where the matching particles of the plasmatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service