Skip to Content
Merck
All Photos(1)

Documents

748927

Sigma-Aldrich

Diethyl carbonate-(carbonyl-13C)

98 atom % 13C, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)213CO
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % 13C

Assay

97% (CP)

form

liquid

bp

126-128 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

CCO[13C](=O)OCC

InChI

1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3/i5+1

InChI key

OIFBSDVPJOWBCH-HOSYLAQJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service