709158
1-Methoxypropane
97%
Synonym(s):
Methyl propyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H10O
CAS Number:
Molecular Weight:
74.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.357
density
0.712 g/mL at 25 °C
functional group
ether
SMILES string
CCCOC
InChI
1S/C4H10O/c1-3-4-5-2/h3-4H2,1-2H3
InChI key
VNKYTQGIUYNRMY-UHFFFAOYSA-N
Application
1-Methoxypropane can be used as:
- A reactant to synthesize organic nitrates via atmospheric oxidation.
- A standard for the calibration curve to quantify thiobarbituric acid reactive substances in proteins.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
<-25.6 °F
Flash Point(C)
<= -32 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yogita Dalla et al.
Indian journal of pharmacology, 41(6), 262-267 (2010-04-22)
High cholesterol levels have been positively correlated with a higher incidence of memory impairment and dementia. The study was undertaken to investigate the potential of the lipid-lowering drug, ezetimibe, in memory deficits associated with dementia of Alzheimer's (AD) type in
Nivedita Sarkar et al.
Journal of applied toxicology : JAT, 38(8), 1071-1080 (2018-03-25)
Arsenite (AsIII) is known for inducing severe oxidative stress and skin carcinogenesis. Contrastingly, phytochemical, epigallocatechin-3-gallate (EGCG) combats toxic insults. Our study focused on the effect of EGCG on redox status of AsIII-stressed normal human keratinocytes, HaCaT cells. EGCG (50 μm) increased
The production of organic nitrates from atmospheric oxidation of ethers and glycol ethers
Espada C and Shepson PB
International Journal of Chemical Kinetics, 37(11), 686-699 (2005)
Elisabetta Bigagli et al.
Journal of photochemistry and photobiology. B, Biology, 173, 618-625 (2017-07-14)
Ultraviolet B (UVB) exposure is a risk factor for corneal damage resulting in oxidative stress, inflammation and cell death. The aim of this study was to investigate the potential protective effects of a commercial eye drop (Dacriovis™) containing Matricaria chamomilla
Dhiman Chakravarty et al.
Plant physiology, 170(2), 761-773 (2015-12-10)
Catalases, enzymes that detoxify H2O2, are widely distributed in all phyla, including cyanobacteria. Unlike the heme-containing catalases, the physiological roles of Mn-catalases remain inadequately characterized. In the cyanobacterium Anabaena, pretreatment of cells with NaCl resulted in unusually enhanced tolerance to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service