Skip to Content
Merck
All Photos(1)

Documents

690104

Sigma-Aldrich

3-(4-Azidophenyl)propionic acid

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Beilstein:
4424312
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

powder

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

OC(=O)CCc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C9H9N3O2/c10-12-11-8-4-1-7(2-5-8)3-6-9(13)14/h1-2,4-5H,3,6H2,(H,13,14)

InChI key

RZYXFEOBOVVBLL-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service