Skip to Content
Merck
All Photos(1)

Documents

678813

Sigma-Aldrich

4-(4-Morpholinyl)benzaldehyde

97%

Synonym(s):

4-Morpholinobenzaldehyde, N-(4-formylphenyl)morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

65-69 °C

SMILES string

[H]C(=O)c1ccc(cc1)N2CCOCC2

InChI

1S/C11H13NO2/c13-9-10-1-3-11(4-2-10)12-5-7-14-8-6-12/h1-4,9H,5-8H2

InChI key

FOAQOAXQMISINY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Murugesan Sankarganesh et al.
Journal of biomolecular structure & dynamics, 39(3), 1055-1067 (2020-02-11)
New monometallic platinum complex ([PtL2Cl2]Cl2) of pyrimidine and morpholine derivative ligand were synthesized and structurally elucidated by elemental analysis, molar conductance, 1H NMR, FT-IR, ESI-MS and UV-Visible spectroscopic techniques. Analytical and spectroscopic result suggests that platinum complex has octahedral geometry.
Ravi Kumar Vuradi et al.
Journal of fluorescence, 26(2), 689-701 (2015-12-29)
New three ruthenium (II) polypyridyl complexes [Ru(phen)2mpip](2+)(1) {mpip = 2-(4-morpholinophenyl)-1H-imidazo[4,5-f][1,10]phenanthroline}, (phen = 1,10-Phenanthrolene), [Ru(bpy)2mpip](2+)(2) (bpy = 2,2'bipyridyl), [Ru(dmb)2mpip](2+)(3) (dmb = 4, 4-dimethyl 2, 2'-bipyridine) have been synthesized and characterized by spectral studies IR, UV-vis, (1)H, (13)C-NMR, mass and elemental analysis. The binding properties of these three complexes towards
Siyang Ding et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-22)
Fluorescent dyes, especially those emitting in the long wavelength region, are excellent candidates in the area of bioassay and bioimaging. In this work, we report a series of simple organic fluorescent dyes consisting of electron-donating aniline groups and electron-withdrawing barbituric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service