606618
Toluene-(phenyl-13C6)
99 atom % 13C
Synonym(s):
Methyl(benzene-13C6)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13C6H5CH3
CAS Number:
Molecular Weight:
98.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Quality Level
Assay
99% (CP)
bp
110 °C (lit.)
mp
-93 °C (lit.)
density
0.920 g/mL at 25 °C
mass shift
M+6
SMILES string
C[13c]1[13cH][13cH][13cH][13cH][13cH]1
InChI
1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i2+1,3+1,4+1,5+1,6+1,7+1
InChI key
YXFVVABEGXRONW-WBJZHHNVSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service