525839
Ammonium niobate(V) oxalate hydrate
99.99% trace metals basis
Synonym(s):
Niobium ammonium oxalate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C4H4NNbO9 · xH2O
Molecular Weight:
302.98 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Recommended Products
grade
for analytical purposes
Assay
99.99% trace metals basis
form
powder
impurities
≤150.0 ppm Trace Metal Analysis
mp
122 °C (dec.) (lit.)
Related Categories
General description
Ammonium niobate(V) oxalate hydrate is a bifunctional reactant that can play the role of niobium precursor and porogenic agent. It is widely used as a starting material to synthesize niobium oxide-based porous nanocomposites, which finds application in the field of photocatalysis, and ceramics .
Application
- Nanostructured Niobium and Titanium Carbonitrides as Electrocatalyst Supports.: This study investigates the synthesis and application of nanostructured niobium and titanium carbonitrides for use as electrocatalyst supports in various electrochemical reactions. The research highlights the potential of these materials to improve the performance and durability of electrocatalysts, making them suitable for applications in fuel cells and other energy conversion devices (McLeod et al., 2024).
- Nb(2)O(5) Coating to Improve the Cyclic Stability and Voltage Decay of Li-Rich Cathode Material for Lithium-Ion Battery.: This paper explores the use of Nb(2)O(5) coatings to enhance the cyclic stability and reduce the voltage decay of lithium-rich cathode materials in lithium-ion batteries. The findings demonstrate that the niobium-based coatings significantly improve the performance and longevity of these battery materials, indicating their utility in advanced battery technologies (Liu et al., 2023).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nb2O5/SiO2 mesoporous monoliths synthetized by sol-gel process using ammonium niobate oxalate hydrate as porogenic agent
Luiz Fernando de Sousa Lima, et al.
Journal of Sol-Gel Science and Technology, 93, 168-174 (2020)
Anchoring dye onto 1D Nb2O5 in cooperation with TEMPO for the selective photocatalytic aerobic oxidation of amines
Xiaoming Ma, et al.
Chemical Engineering Journal, 426, 131418-131418 (2021)
Phase transition behaviour and physicochemical properties of KNbO3 ceramics
P. Vlazan, et al.
Ceramics International, 43, 5963-5967 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service