Skip to Content
Merck
All Photos(1)

Key Documents

444197

Sigma-Aldrich

3-Ethyl-3-oxetanemethanol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.453 (lit.)

bp

96 °C/4 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

CCC1(CO)COC1

InChI

1S/C6H12O2/c1-2-6(3-7)4-8-5-6/h7H,2-5H2,1H3

InChI key

UNMJLQGKEDTEKJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of fluorescent hyperbranched polyether.
Gao C, et al.
Reactive functional Polymers, 58(1), 65-72 (2004)
Paweł Parzuchowski et al.
Polymers, 12(1) (2020-01-23)
Hyperbranched polyoxetanes are a relatively new class of polymers. These are branched polyethers that are synthesized from oxetanes-four-member cyclic ethers bearing hydroxymethyl groups-via ring-opening polymerization. Four series of polyoxetanes were synthesized from 3-ethyl-3-(hydroxymethyl)oxetane and 1,1,1-tris(hydroxymethyl)propane as a core molecule. Reagents
Qi Zhu et al.
The journal of physical chemistry. B, 113(17), 5777-5780 (2009-04-03)
The influence of branching architecture on the glass transition of hyperbranched polyethers has been investigated. For amorphous samples, the glass transition temperature (T(g)) first increases with the degree of branching (DB), passes through a maximum, and then decreases sharply. An
A study of monomer's effect on adhesion strength of UV-curable resins.
Chiang TH and Hsieh T-E.
International Journal of Adhesion and Adhesives, 26(7), 520-531 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service