Skip to Content
Merck
All Photos(1)

Key Documents

381896

Sigma-Aldrich

2,3-Naphthalocyanine

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H26N8
CAS Number:
Molecular Weight:
714.77
Beilstein:
4345876
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 95%

Quality Level

λmax

712 nm

SMILES string

c1ccc2cc3c4nc(nc5[nH]c(nc6nc(nc7[nH]c(n4)c8cc9ccccc9cc78)c%10cc%11ccccc%11cc6%10)c%12cc%13ccccc%13cc5%12)c3cc2c1

InChI

1S/C48H26N8/c1-2-10-26-18-34-33(17-25(26)9-1)41-49-42(34)54-44-37-21-29-13-5-6-14-30(29)22-38(37)46(51-44)56-48-40-24-32-16-8-7-15-31(32)23-39(40)47(52-48)55-45-36-20-28-12-4-3-11-27(28)19-35(36)43(50-45)53-41/h1-24H,(H2,49,50,51,52,53,54,55,56)

InChI key

LKKPNUDVOYAOBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoning Li et al.
Theranostics, 8(3), 767-784 (2018-01-19)
Fluorescence image-guided surgery combined with intraoperative therapeutic modalities has great potential for intraoperative detection of oncologic targets and eradication of unresectable cancer residues. Therefore, we have developed an activatable theranostic nanoplatform that can be used concurrently for two purposes: (1)
Yixiao Guo et al.
ACS applied materials & interfaces, 10(24), 20884-20896 (2018-06-13)
Brown adipose tissue (BAT) has been identified as a promising target for the treatment of obesity, diabetes, and relevant metabolism disorders because of the adaptive thermogenesis ability of this tissue. Visualizing BAT may provide an essential tool for pathology study
Abraham S Moses et al.
Small (Weinheim an der Bergstrasse, Germany), 16(18), e1906936-e1906936 (2020-04-07)
Endometriosis is a painful disorder where endometrium-like tissue forms lesions outside of the uterine cavity. Intraoperative identification and removal of these lesions are difficult. This study presents a nanoplatform that concurrently delineates and ablates endometriosis tissues using real-time near-infrared (NIR)
V Mantareva et al.
Journal of photochemistry and photobiology. B, Biology, 40(3), 258-262 (1998-02-12)
A Si(IV)-naphthalocyanine bearing two methoxyethylenglycol axial ligands to the centrally coordinated metal ion (SiNc) was prepared by chemical synthesis and assayed for the phototherapeutic activity after administration in a Cremophor formulation to C57BI/6 mice bearing a subcutaneously transplanted Lewis lung
N Michailov et al.
Journal of photochemistry and photobiology. B, Biology, 37(1-2), 154-157 (1997-01-01)
In vivo experiments were performed to evaluate the effect of fluence rate on the efficiency of Zn(II)-2,3 naphthalocyanine (ZnNc) photosensitization of B16 pigmented melanoma subcutaneously transplanted in C57B1/6 mice. The tumour was irradiated with 774 nm light at 24 h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service