Skip to Content
Merck
All Photos(2)

Documents

371114

Sigma-Aldrich

1-Deoxy-1-(octylamino)-D-glucitol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H31NO5
CAS Number:
Molecular Weight:
293.40
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]20/D −15°, c = 1 in methanol

mp

121-124 °C (lit.)

SMILES string

CCCCCCCCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C14H31NO5/c1-2-3-4-5-6-7-8-15-9-11(17)13(19)14(20)12(18)10-16/h11-20H,2-10H2,1H3/t11-,12+,13+,14+/m0/s1

InChI key

ZRRNJJURLBXWLL-REWJHTLYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service