Skip to Content
Merck
All Photos(1)

Key Documents

367478

Sigma-Aldrich

O-(tert-Butyldimethylsilyl)hydroxylamine

95%

Synonym(s):

Aminoxy-tert-butyldimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2ONH2
CAS Number:
Molecular Weight:
147.29
Beilstein:
2424306
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

87-90 °C/40 mmHg (lit.)

mp

62-65 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)ON

InChI

1S/C6H17NOSi/c1-6(2,3)9(4,5)8-7/h7H2,1-5H3

InChI key

SSUCKKNRCOFUPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine) may be used to induce the cleavage, during solid supported synthesis of hydroxamic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid supported synthesis of hydroxamic acids.
Golebiowski A and Klopfenstein S.
Tetrahedron Letters, 39(21), 3397-3400 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service