Skip to Content
Merck
All Photos(2)

Key Documents

359939

Sigma-Aldrich

(R)-(+)-Bornylamine

97%

Synonym(s):

(+)-Bornanamine, endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N
CAS Number:
Molecular Weight:
153.26
Beilstein:
2801831
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +44°, c = 0.5 in ethanol

mp

160-163 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)[C@@H](N)C2

InChI

1S/C10H19N/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8H,4-6,11H2,1-3H3/t7-,8+,10+/m1/s1

InChI key

MDFWXZBEVCOVIO-WEDXCCLWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-Bornylamine in the presence of acetonitrile as a solvent and tetraethylammonium bromide as the bromide ion source, may be used in the crystallization-induced chiral inversion of (S)-2-bromo-3-phenylpropanoic acid to its (R)-enantiomer. It may also be used to synthesize trans-bis{(R)-(+)-bornylamino}palladium(II) dichloride, a palladium(II) complex with potent antitumor activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization-Induced Chiral Inversion As the Key Step for Synthesis of (S)-2-Acetylthio-3-phenylpropanoic Acid from l-Phenylalanine.
Chen JG, et al.
Organic Letters, 6(19), 3233-3235 (2004)
Synthesis, crystal structure and initial biological evaluation of the new enantiomerically pure chiral palladium (II) complex trans-bis {endo-(1R)-1, 7, 7-trimethylbicyclo [2.2. 1]-heptan-2-amino} palladium (II) dichloride.
Abu-Surrah AS, et al.
European Journal of Medicinal Chemistry, 37(11), 919-922 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service