359300
Sodium 2-methyl-2-propanethiolate
technical grade, 90%
Synonym(s):
2-Methyl-2-propanethiol sodium salt, Sodium tert-butyl sulfide, t-Butanethiol sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CSNa
CAS Number:
Molecular Weight:
112.17
Beilstein:
3682306
MDL number:
UNSPSC Code:
12352105
PubChem Substance ID:
NACRES:
NA.07
Assay:
90%
grade:
technical grade
form:
crystalline powder
powder, crystals or chunks
powder, crystals or chunks
Recommended Products
grade
technical grade
Quality Level
Assay
90%
form
crystalline powder
powder, crystals or chunks
SMILES string
CC(C)(C)S[Na]
InChI
1S/C4H10S.Na/c1-4(2,3)5;/h5H,1-3H3;/q;+1/p-1
InChI key
TUZFIWBNGKNFPW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Exploration of 9-bromo [7] helicene reactivity.
Zadny J, et al.
Tetrahedron, 69(3), 6213-6218 (2013)
Shih-Ching Chuang et al.
The Journal of organic chemistry, 72(17), 6447-6453 (2007-07-20)
In the field of open-cage fullerenes, there was a lack of a universal standard that could correlate and quantify the orifice size of open-cage fullerenes. One cannot compare the relative orifice size by simple comparison of the number of atoms
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service