Skip to Content
Merck
All Photos(3)

Documents

345865

Sigma-Aldrich

Ammonium trifluoromethanesulfonate

99%

Synonym(s):

Ammonium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3NH4
CAS Number:
Molecular Weight:
167.11
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

crystals and lumps

mp

224-226 °C (lit.)

SMILES string

N.OS(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.H3N/c2-1(3,4)8(5,6)7;/h(H,5,6,7);1H3

InChI key

BMWDUGHMODRTLU-UHFFFAOYSA-N

General description

Ammonium trifluoromethanesulfonate is a white crystalline solid that is highly soluble in water. It is thermally stable and widely used in the field of catalysis, batteries, and organic synthesis.

Application

Ammonium trifluoromethanesulfonate can be used:
  • As an electrolyte additive in aqueous metal ion batteries. It is used to enhance the ionic conductivity of electrolyte and widens the electrochemical stability window of the battery.
  • As a precursor to prepare solid polymer electrolytes and gel polymer electrolytes via solution casting by method.
  • As a co-catalyst to prepare Poly(p-phenylenediamine) and their derivatives. These conductive polymers are used in sensors and organic photovoltaic cells.
  • A dopant in the preparation of flexible poly(vinylidene fluoride-co-hexafluoropropylene)/polyethyl methacrylate ammonium triflate (PVDf-HFP/PEMA-NH4CF3SO3)polymer electrolyte films.
  • A cathode additive in proton exchange membrane (PEM) fuel cells to increase the local oxygen concentration near the Pt catalyst, which enhances single-cell performance.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sultan Ahmed et al.
Nanotechnology, 29(39), 395401-395401 (2018-07-04)
Research and development on all-solid-state, flexible supercapacitors is the prime concern of the scientific community these days due to their various advantages including their easy transportability, miniaturization, and compactness in different appliances. We report the novel configuration of all-solid symmetrical
Russell J Needham et al.
Journal of inorganic biochemistry, 210, 111154-111154 (2020-08-11)
Twenty-four novel organometallic osmium(II) phenylazopyridine (AZPY) complexes have been synthesised and characterised; [Os(η6-arene)(5-RO-AZPY)X]Y, where arene = p-cym or bip, AZPY is functionalized with an alkoxyl (O-R, R = Me, Et, nPr, iPr, nBu) or glycolic (O-{CH2CH2O}nR*, n = 1-4, R* = H, Me, or Et) substituent on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service