Skip to Content
Merck
All Photos(4)

Documents

309338

Sigma-Aldrich

Nickel(II) perchlorate hexahydrate

Synonym(s):

Nickel(2+) diperchlorate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(ClO4)2 · 6H2O
CAS Number:
Molecular Weight:
365.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

crystalline

Quality Level

reaction suitability

reagent type: oxidant

concentration

15.3-17.0% Ni (EDTA titration)

mp

140 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.[Ni++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Ni.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2

InChI key

WHGYCGOFTBFDLW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 3 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy C. Higgs et al.
Inorganic chemistry, 37(9), 2263-2272 (2001-10-24)
The mixed functionality pyrazole/phenol ligand (2-hydroxyphenyl)bis(pyrazolyl)methane, L1OH, has been used to prepare a series of linear trimetallic systems with the general structural motif [M(3)(L1O)(4)](2+), where M = Mn(2+), Co(2+), Ni(2+), Cu(2+), and Zn(2+). Each of these complexes has been structurally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service