Skip to Content
Merck
All Photos(1)

Key Documents

287997

Sigma-Aldrich

1,5-Bis(diphenylphosphino)pentane

97%

Synonym(s):

1,5-pentanediylbis[diphenylphosphine], DPPENT, Pentamethylenebis[diphenylphosphine], dpppe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2P(CH2)5P(C6H5)2
CAS Number:
Molecular Weight:
440.50
Beilstein:
2953640
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

43-47 °C (lit.)

functional group

phosphine

SMILES string

C(CCP(c1ccccc1)c2ccccc2)CCP(c3ccccc3)c4ccccc4

InChI

1S/C29H30P2/c1-6-16-26(17-7-1)30(27-18-8-2-9-19-27)24-14-5-15-25-31(28-20-10-3-11-21-28)29-22-12-4-13-23-29/h1-4,6-13,16-23H,5,14-15,24-25H2

InChI key

MZFPAWGWFDGCHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Zain Aldin et al.
Dalton transactions (Cambridge, England : 2003), 47(39), 13926-13938 (2018-09-19)
The reaction of [RuCl2(p-cymene)]2 with potassium O-ethylxanthate and a set of nine representative Ph2P-X-PPh2 bidentate phosphines (dppm, dppe, dppp, dppb, dpppe, dppen, dppbz, dppf, and DPEphos) afforded monometallic [Ru(S2COEt)2(diphos)] chelates 1-9 in 62-96% yield. All the products were fully characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service