Skip to Content
Merck
All Photos(1)

Key Documents

257621

Sigma-Aldrich

2,2-Di(4-tert-octylphenyl)-1-picrylhydrazyl, free radical

Synonym(s):

2,2-Bis[4-(1,1,3,3-tetramethylbutyl)phenyl]-1-(2,4,6-trinitrophenyl)hydrazinyl, 2,2-Di(4-tert-octylphenyl)-1-picrylhydrazyl, DPPH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H44N5O6
CAS Number:
Molecular Weight:
618.74
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

171 °C (dec.) (lit.)

λmax

531 nm

functional group

nitro

storage temp.

2-8°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(cc1)N([N]c2c(cc(cc2[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)c3ccc(cc3)C(C)(C)CC(C)(C)C

InChI

1S/C34H44N5O6/c1-31(2,3)21-33(7,8)23-11-15-25(16-12-23)36(26-17-13-24(14-18-26)34(9,10)22-32(4,5)6)35-30-28(38(42)43)19-27(37(40)41)20-29(30)39(44)45/h11-20H,21-22H2,1-10H3

InChI key

CLOCMRDQPYXVKP-UHFFFAOYSA-N

General description

2,2-Di(4-tert-octylphenyl)-1-picrylhydrazyl (DPPH) is classified as a “stable” free radical since it has a long shelf life and can be synthesized and stored under normal laboratory conditions.

Application

2,2-Di(4-tert-octylphenyl)-1-picrylhydrazyl can be used as a calibration standard in electron paramagnetic spectroscopy (EPR) and as a radical scavenger in polymers. It is also used as an efficient reagent in the “DPPH calorimetric assay” to study radical scavenging activities of diverse chemical compounds, mostly of phenolic nature. When DDPF reacts with antioxidant molecules, its color changes from deep purple to pale yellow, which allows the spectrophotometric determination of the antioxidant activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DPPH (2, 2-di (4-tert-octylphenyl)-1-picrylhydrazyl) radical scavenging mixed-mode colorimetric assay (s)
Measurement of Antioxidant Activity and Capacity: Recent Trends and Applications, 141-164 (2017)
Zeynep Akar et al.
Journal of enzyme inhibition and medicinal chemistry, 32(1), 640-647 (2017-03-07)
2,2-Diphenyl-1-picrylhydrazyl (DPPH•) radical scavenging, the most commonly used antioxidant method with more than seventeen thousand articles cited, is very practical; however, as with most assays, it has the major disadvantage of dependence on a spectrophotometer. To overcome this drawback, the
Ramona Figat et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-04)
Traditional medicinal plants are an important source of active compounds with potential antimutagenic activity. Polyscias filicifolia Bailey (Araliaceae) is a South Asian traditional herb used as an adaptogenic and cardiac drug. Extracts of P. filicifolia contain a wide range of
Huanhuan Xu et al.
Frontiers in cell and developmental biology, 8, 822-822 (2020-10-06)
Caffeine is chemically stable and not readily oxidized under normal physiological conditions but also has antioxidant effects, although the underlying molecular mechanism is not well understood. Superoxide dismutase (SOD) 2 is a manganese-containing enzyme located in mitochondria that protects cells
Christian Galasso et al.
International journal of molecular sciences, 21(22) (2020-11-22)
The antioxidant activity of natural compounds consists in their ability to modulate gene and protein expression, thus inducing an integrated cell protective response and repair processes against oxidative stress. New screening tools and methodologies are crucial for the actual requirement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service