Skip to Content
Merck
All Photos(1)

Documents

251720

Sigma-Aldrich

Pentaerythritol allyl ether

technical grade, 70%

Synonym(s):

Allyl pentaerythritol, Pentaerythritol triallyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2OCH2CH=CH2)x(CH2OH)y
CAS Number:
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

Assay

70%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

100 °C/0.1 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

OCC(COCC=C)(COCC=C)COCC=C

InChI

1S/C8H16O4/c1-2-3-12-7-8(4-9,5-10)6-11/h2,9-11H,1,3-7H2

InChI key

RFIMISVNSAUMBU-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service