Skip to Content
Merck
All Photos(1)

Key Documents

223271

Sigma-Aldrich

Trifluorothymine

97%

Synonym(s):

5-Trifluoromethyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2O2
CAS Number:
Molecular Weight:
180.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

245-246 °C (lit.)

solubility

methanol: soluble 50 mg/mL, clear

functional group

fluoro

SMILES string

FC(F)(F)C1=CNC(=O)NC1=O

InChI

1S/C5H3F3N2O2/c6-5(7,8)2-1-9-4(12)10-3(2)11/h1H,(H2,9,10,11,12)

InChI key

LMNPKIOZMGYQIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Salvetti et al.
Nucleosides, nucleotides & nucleic acids, 20(4-7), 1123-1125 (2001-09-21)
Recently, beta-L-nucleoside analogues have emerged as a new class of sugar modified nucleosides with potential antiviral and/or antitumoral activity. As a part of our ongoing research on this topic, we decided to synthesize 5-CF3-beta-L-dUrd (7), the hitherto unknown L-enantiomer of
R Pouremad et al.
NMR in biomedicine, 12(6), 373-380 (1999-10-12)
Metabolism of trifluorothymidine (TFT) and its transport across the blood-brain barrier (BBB) has been measured quantitatively in rats by fluorine-19 nuclear magnetic resonance spectroscopy ((19)F NMR). It is demonstrated that TFT crosses the BBB in micromolar quantities and is metabolized
Cağrı Cırak et al.
Journal of molecular modeling, 18(9), 4453-4464 (2012-05-31)
In the present work, the experimental and the theoretical vibrational spectra of trifluorothymine were investigated. The FT-IR (400-4000 cm(-1)) and μ-Raman spectra (100-4000 cm(-1)) of trifluorothymine in the solid phase were recorded. The geometric parameters (bond lengths and bond angles) and vibrational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service