Skip to Content
Merck
All Photos(1)

Key Documents

220566

Sigma-Aldrich

(S)-(−)-α-Methylbenzyl isocyanate

98%

Synonym(s):

(S)-(−)-1-Phenylethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NCO
CAS Number:
Molecular Weight:
147.17
Beilstein:
4230972
EC Number:
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −10°, neat

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.5145 (lit.)

bp

55-56 °C/2.5 mmHg (lit.)

density

1.045 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](N=C=O)c1ccccc1

InChI

1S/C9H9NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-6,8H,1H3/t8-/m0/s1

InChI key

JJSCUXAFAJEQGB-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-α-Methylbenzyl isocyanate is a commonly used chiral derivatizing agent for the resolution of enantiomers.

Application

(S)-(-)-α-Methylbenzyl isocyanate may be used as a chiral auxiliary for the resolution of racemic (±)-6aR,11aR-maackiain to give the (-)-form, which is a key intermediate for synthesizing (-)-cabenegrin A-I.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Absolute configuration and total synthesis of (-)-cabenegrin AI.
Tokes AL, et al.
Tetrahedron, 55(30), 9283-9296 (1999)
Resolution of salbutamol enantiomers in human urine by reversed-phase high performance liquid chromatography after derivatization with (S)-(-)-a-methylbenzyl isocyanate.
Kim KH, et al.
Archives of Pharmacal Research, 20(5), 486-490 (1997)
Determination of the optical purity of (R)-terbutaline by 1 H-NMR and RP-LC using chiral derivatizing agent,(S)-(-)-a-methylbenzyl isocyanate.
Kim KH, et al.
Journal of Pharmaceutical and Biomedical Analysis, 25(5), 947-956 (2001)
Determination of pindolol enantiomers in human plasma and urine by simple liquid-liquid extraction and high-performance liquid chromatography.
Beal JL and Tett SE
Journal of Chromatography. B, Biomedical Applications, 715(2), 409-415 (1998)
High-performance liquid chromatographic determination of the enantiomers of beta-adrenoceptor blocking agents in biological fluids. II. Studies with atenolol.
S K Chin et al.
Journal of chromatography, 489(2), 438-445 (1989-04-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service