Skip to Content
Merck
All Photos(1)

Key Documents

216585

Supelco

Amberlite IRA-900 Ion Exchange Resin

chloride form, 16-50 mesh

Synonym(s):

mix mode ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NB.21

Product Name

Amberlite IRA-900 chloride form, chloride form

vapor density

<1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

form

beads

crosslinking

>95 % cross-linked

autoignition temp.

800 °F

parameter

60 °C OH- form max. temp.
77 °C Cl- form max. temp.

moisture

58-64%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

quaternary ammonium

particle size

650-820 μm

operating pH

0-14

capacity

1.0 meq/mL by wetted bed volume

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite IRA-900 chloride form is a macroporous, polystyrenic type 1 resin. This resin was found to catalyze the photodecomposition of carbon tetrachloride in ethanol.

Application

Amberlite IRA-900 modified with metallophthalocyanines was used to remove 4-nitorphenol.
Strongly basic (type I) anion exchange resin for decolorizing intermediate to light solutions, concentrating heparin at neutral pH, removing metal ions, etc.

Features and Benefits

Strongly basic, macroreticular resin of moderately high porosity with benzyltrialkylammonium functionality which is useful in decolorizing and removal of organic materials.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eloïse Marais et al.
Journal of hazardous materials, 152(1), 293-301 (2007-08-08)
The adsorption of 4-nitrophenol using commercially available Amberlite IRA-900 modified with metal phthalocyanines (MPc) was investigated. The metallophthalocyanines immobilised onto the surface of Amberlite IRA-900 include Fe (FePcS4), Co (CoPcS4) and Ni (NiPcS4) tetrasulphophthalocyanines, and differently sulphonated phthalocyanine mixtures of
Catalysis of the photodecomposition of carbon tetrachloride in ethanol by an amberlite anion exchange resin.
Hoggard, Patrick E., and Andrea Maldotti.
J. Catal., 275, 243-249 (2010)
Comparison of strongly basic anion exchange resins applicability for the removal of palladium (II) ions from acidic solutions
Wolowicz, A., and Z. Hubicki
Chemical Engineering Journal, 171, 206-215 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service