Skip to Content
Merck
All Photos(2)

Documents

185396

Sigma-Aldrich

Tetrahydrofurfuryl alcohol

99%

Synonym(s):

NSC 15434, Tetrahydro-2-furanmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Beilstein:
102723
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39150702
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.52 (vs air)

Quality Level

vapor pressure

2.3 mmHg ( 39 °C)

Assay

99%

form

liquid

expl. lim.

9.7 %

refractive index

n20/D 1.452 (lit.)

bp

178 °C (lit.)

mp

−80 °C (lit.)

solubility

acetone: miscible(lit.)
alcohol: miscible(lit.)
benzene: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: miscible(lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCO1

InChI

1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2

InChI key

BSYVTEYKTMYBMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofurfuryl alcohol undergoes chemoselective hydrogenolysis catalyzed by Rh/SiO2 modified with ReOx species to yield 1,5-pentanediol. It undergoes lanthanum-mediated Michael-type addition reaction with maleate to form alkoxybutanedioic acid.

Application

Tetrahydrofurfuryl alcohol was used as refractive-index matching solvent to ensure hard sphere suspension of silica particles for rheological measurements.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microstructure and rheological behaviour of electrosterically stabilized silica particle suspensions.
So J-H, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 190(1), 89-98 (2001)
LaIII-lnduced Addition of Tetrahydrofurfuryl Alcohol, Tetrahydropyran-2-ylmethanol, D-Gluconate, Methanol and Ethanol to Maleate.
Quartey EGK, et al.
Acta Chemica Scandinavica, 50, 825-831 (1996)
T Schräder et al.
Journal of bacteriology, 183(24), 7408-7411 (2001-11-22)
Different aldehyde dehydrogenases (AlDHs) were formed during growth of Ralstonia eutropha Bo on tetrahydrofurfuryl alcohol (THFA). One of these enzymes, AlDH 4, was purified and characterized as a homodimer containing no prosthetic groups, showing a strong substrate inhibition, and having
Chung-Yi Chen
Natural product research, 24(19), 1830-1833 (2010-11-26)
Pressalanin A, a 2-functionalized tetrahydrofuranol, has been isolated from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae), and the structure was determined on the basis of a spectroscopic analysis.
S M Bellm et al.
The Journal of chemical physics, 136(24), 244301-244301 (2012-07-05)
Cross section data for electron scattering from DNA are important for modelling radiation damage in biological systems. Triply differential cross sections for the electron impact ionization of the highest occupied outer valence orbital of tetrahydrofurfuryl alcohol, which can be considered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service