Skip to Content
Merck
All Photos(1)

Key Documents

17782

Sigma-Aldrich

Triglycerol

technical

Synonym(s):

1,3-Bis(2,3-dihydroxypropyl)-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH(CH2OCH2CHOHCH2OH)2
CAS Number:
Molecular Weight:
240.25
Beilstein:
1724207
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

impurities

≤20% Di- and Tetraglycerol

density

1.30 g/mL at 20 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OCC(O)COCC(O)COCC(O)CO

InChI

1S/C9H20O7/c10-1-7(12)3-15-5-9(14)6-16-4-8(13)2-11/h7-14H,1-6H2

InChI key

AGNTUZCMJBTHOG-UHFFFAOYSA-N

Application

Triglycerol was used in the synthesis of (poly)glycerol monoethers. It was also used to enhance the chromatographic behavior of the pesticides in tea matrices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Nešić et al.
Carbohydrate polymers, 157, 981-990 (2016-12-19)
In this study, pectin based films including different amounts of sodium alginate were prepared by casting method. All the films, with and without polyglycerol as plasticizer, were crosslinked with zinc ions in order to extend their potential functionality. The development
Tomiyasu Murata et al.
FEBS open bio, 10(10), 1967-1984 (2020-08-13)
Dysregulation of adipocyte differentiation and dysfunction play key roles in the pathogenesis of obesity and associated disorders such as diabetes and metabolic syndrome, and as such, a better understanding of the molecular mechanism of adipogenesis may help to elucidate the
Christina Yfanti et al.
Journal of sports science & medicine, 16(3), 375-382 (2017-09-16)
Eccentric exercise has been shown to exert beneficial effects in both lipid profile and insulin sensitivity. Antioxidant supplementation during chronic exercise is controversial as it may prevent the physiological training-induced adaptations. The aim of this study was to investigate: 1)
Nurul Elyani Mohamad et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 1257962-1257962 (2020-10-09)
Obesity is a pandemic metabolic syndrome with increasing incidences every year. Among the significant factors that lead to obesity, overconsumption of high-fat food in daily intake is always the main contributor. Functional foods have shown a positive effect on disease
Cecilia Casali et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158616-158616 (2020-01-14)
In renal cells, hyperosmolarity can induce cellular stress or differentiation. Both processes require active endoplasmic reticulum (ER)-associated protein synthesis. Lipid biosynthesis also occurs at ER surface. We showed that hyperosmolarity upregulates glycerophospholipid (GP) and triacylglycerol (GL-TG) de novo synthesis. Considering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service