Skip to Content
Merck
All Photos(1)

Documents

15469

Sigma-Aldrich

Boc-Homophe-OH

≥98.0% (TLC)

Synonym(s):

(S)-2-(Boc-amino)-4-phenylbutyric acid, Boc-L-homophenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH[NHCOOC(CH3)3]COOH
CAS Number:
Molecular Weight:
279.33
Beilstein:
3653506
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]20/D +6.5±1°, c = 2% in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

76-80 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCc1ccccc1)C(O)=O

InChI

1S/C15H21NO4/c1-15(2,3)20-14(19)16-12(13(17)18)10-9-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,16,19)(H,17,18)/t12-/m0/s1

InChI key

MCODLPJUFHPVQP-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Voegeli et al.
International journal of cosmetic science, 39(2), 109-120 (2016-07-20)
The aim of this study was to optimize the synthesis of the plasmin and urokinase (uPA) inhibitor benzylsulfonyl-D-Ser-homoPhe-(4-amidino-benzylamide) (BSFAB), to characterize its activity and mechanism of action and to assess its use to improve stratum corneum (SC) barrier function. Peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service