All Photos(1)
About This Item
Linear Formula:
(CH3O)3C6H2CHO
CAS Number:
Molecular Weight:
196.20
Beilstein:
981091
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
refractive index
n20/D 1.5547 (lit.)
bp
168-170 °C/12 mmHg (lit.)
mp
38-40 °C (lit.)
SMILES string
[H]C(=O)c1ccc(OC)c(OC)c1OC
InChI
1S/C10H12O4/c1-12-8-5-4-7(6-11)9(13-2)10(8)14-3/h4-6H,1-3H3
InChI key
UCTUXUGXIFRVGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2,3,4-Trimethoxybenzaldehyde was used to study the effects of 2,3,4-trimethoxybenzaldehyde on tubulin-dependent GTP hydrolysis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
C M Lin et al.
The Journal of biological chemistry, 256(17), 9242-9245 (1981-09-10)
The effects of a number of antimitotic drugs on the GTPase activity of tubulin were examined. The previously reported stimulation with colchicine and inhibition with podophyllotoxin and vinblastine wee confirmed. Maytansine, which competes with vinblastine in binding to tubulin, was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service