Skip to Content
Merck
All Photos(1)

Key Documents

15052

Sigma-Aldrich

Boc-Ala-ONp

≥96.0% (HPLC)

Synonym(s):

Boc-L-alanine 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O6
CAS Number:
Molecular Weight:
310.30
Beilstein:
1892073
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C[C@H](NC(=O)OC(C)(C)C)C(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C14H18N2O6/c1-9(15-13(18)22-14(2,3)4)12(17)21-11-7-5-10(6-8-11)16(19)20/h5-9H,1-4H3,(H,15,18)/t9-/m0/s1

InChI key

SUHFNHHZORGDFI-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Sohl et al.
Biochemistry, 36(13), 3894-3902 (1997-04-01)
alpha-Lytic protease, a chymotrypsin-like serine protease, is synthesized with an N-terminal 166 amino acid pro region which is absolutely required for folding of the protease. The pro region is also the most potent inhibitor of the protease known with a
H M Korchak et al.
The Journal of biological chemistry, 259(12), 7439-7445 (1984-06-25)
Activated neutrophils aggregate, generate superoxide (O-2), and degranulate. The role of Ca as "second messenger" in neutrophil activation was examined using as agonist the chemotactic peptide fMet-Leu-Phe and its antagonist t-butoxycarbonyl-Phe-Leu-Phe-Leu-Phe to systematically vary the time of receptor occupancy. Release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service