Skip to Content
Merck
All Photos(1)

Key Documents

122327

Sigma-Aldrich

Quinoline N-oxide hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO · xH2O
CAS Number:
Molecular Weight:
145.16 (anhydrous basis)
Beilstein:
5989589
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

52-55 °C (lit.)

SMILES string

[H]O[H].[O-][n+]1cccc2ccccc12

InChI

1S/C9H7NO.H2O/c11-10-7-3-5-8-4-1-2-6-9(8)10;/h1-7H;1H2

InChI key

CUSWDTBBCIXCRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinoline N-oxide hydrate forms complexes with lanthanide chloride.

Application

Quinoline N-oxide hydrate was used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The application of trifluoroacetic anhydride-sodium iodide (TFAA-I) system for quantitative determination of nitrones.
Clesielski W, et al.
Canadian Journal of Chemistry, 68(5), 679-684 (1990)
Lanthanide chloride complexes with quinoline-n-oxide.
Kingston JV, et al.
J. Inorg. Nucl. Chem., 31(10), 3181-3185 (1969)
Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service