Skip to Content
Merck
All Photos(2)

Key Documents

120251

Sigma-Aldrich

Diethyl 2,6-dimethylpyridine-3,5-dicarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO4
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

208 °C/40 mmHg (lit.)

mp

72-74 °C (lit.)

SMILES string

CCOC(=O)c1cc(C(=O)OCC)c(C)nc1C

InChI

1S/C13H17NO4/c1-5-17-12(15)10-7-11(13(16)18-6-2)9(4)14-8(10)3/h7H,5-6H2,1-4H3

InChI key

DIIWSYPKAJVXBV-UHFFFAOYSA-N

General description

Diethyl 2,6-dimethylpyridine-3,5-dicarboxylate is formed during the oxidation of 4-substituted Hantsch dihydropyridines in presence of methanesulfonic acid, sodium nitrite and wet SiO2 as oxidizing agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service