Skip to Content
Merck
All Photos(1)

Documents

113247

Sigma-Aldrich

1-Phenyltridecane

99%

Synonym(s):

Tridecylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)12CH3
CAS Number:
Molecular Weight:
260.46
Beilstein:
1947201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.482 (lit.)

mp

10 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCc1ccccc1

InChI

1S/C19H32/c1-2-3-4-5-6-7-8-9-10-11-13-16-19-17-14-12-15-18-19/h12,14-15,17-18H,2-11,13,16H2,1H3

InChI key

MCVUKOYZUCWLQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-phenyltridecane was converted to trans-cinnamic and 3-phenylpropionic acid by an oil-degrading bacterium.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>233.6 °F - closed cup

Flash Point(C)

> 112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O O Amund et al.
Antonie van Leeuwenhoek, 51(1), 45-56 (1985-01-01)
An oil-degrading bacterium identified as Acinetobacter lwoffi was isolated by elective culture on North Sea Forties crude oil from an activated sludge sample. It grew on a wide range of n-alkanes (C12-C28) and 1-phenylalkanes, including 1-phenyldodecane, 1-phenyltridecane and 1-phenyltetradecane. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service