Skip to Content
Merck
All Photos(1)

Key Documents

102288

Sigma-Aldrich

2′-Hydroxy-3-phenylpropiophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH2CH2C6H5
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

refractive index

n20/D 1.5968 (lit.)

mp

36-37 °C (lit.)

functional group

ketone
phenyl

SMILES string

Oc1ccccc1C(=O)CCc2ccccc2

InChI

1S/C15H14O2/c16-14-9-5-4-8-13(14)15(17)11-10-12-6-2-1-3-7-12/h1-9,16H,10-11H2

InChI key

JCPGMXJLFWGRMZ-UHFFFAOYSA-N

General description

2′-Hydroxy-3-phenylpropiophenone is a colorless to yellow liquid or solid. 2′-Hydroxy-3-phenylpropiophenone derivatives show antiarrhythmic,spasmolytic and local anaesthetic activities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Local anesthetic activity of derivatives of 2-hydroxy-3-phenylpropiophenone.
Nikolaevskii VA, et al.
Pharmaceutical Chemistry Journal, 21(9), 646-647 (1987)
Antiarrhythmic activity of 2'-hydroxy-3-phenylpropiophenone derivatives.
Nikolaevskii VA, et al.
Pharmaceutical Chemistry Journal, 23(12), 966-968 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service