Merck
Bio-electro oxidation of indigo carmine by using microporous activated carbon fiber felt as anode and bioreactor support.
Chemosphere (2017-08-16)
Luane Ferreira Garcia, Ana Claudia Rodrigues Siqueira, Germán Sanz Lobón, Jossano Saldanha Marcuzzo, Benevides Costa Pessela, Eduardo Mendez, Telma Alves Garcia, Eric de Souza Gil
RÉSUMÉ
The bioremediation and electro-oxidation (EO) processes are included among the most promising cleaning and decontamination mechanisms of water. The efficiency of bioremediation is dictated by the biological actuator for a specific substrate, its suitable immobilization and all involved biochemical concepts. The EO performance is defined by the anode efficiency to perform the complete mineralization of target compounds and is highlighted by the low or null use of reagent. Recently, the combination of both technologies has been proposed. Thus, the development of high efficient, low cost and eco-friendly anodes for sustainable EO, as well as, supporting devices for immobilization of biological systems applied in bioremediation is an open field of research. Therefore, the aim of this work was to promote the bio-electrochemical remediation of indigo carmine dye (widely common in textile industry), using new anode based on a microporous activated carbon fiber felt (ACFF) and ACFF with immobilized Laccase (Lcc) from Pycnoporus sanguineus. The results were discolorations of 62.7% with ACFF anode and 83.60% with ACFF-MANAE-Lcc anode, both for 60 min in tap water. This remediation rates show that this new anode has low cost and efficiency in the degradation of indigo dye and can be applied for other organic pollutant.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2,5-Dimethylaniline, 99%
Le tarif et la disponibilité ne sont pas disponibles actuellement.