Merck
Resistance irrelevant CYP417A2v2 was found degrading insecticide in
Ecology and evolution (2017-08-05)
Mohammad Asaduzzaman Miah, Mohammed Esmail Abdalla Elzaki, Zhaojun Han
RÉSUMÉ
Cytochrome P450 monooxygenases (CYPs) usually overexpressed in resistant strain were found involved in oxidative detoxification of insecticides. In this study, an investigation was conducted to confirm if resistance irrelevant CYPs which were not overexpressed in resistant strain before, were capable of degrading insecticides. Three resistance irrelevant CYPs viz. CYP417A2v2, CYP425A1v2, and CYP4DJ1 from CYP4 family of
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Nitroanisole, ≥99%