- Preferences for phosphorylation sites in the retinoblastoma protein of D-type cyclin-dependent kinases, Cdk4 and Cdk6, in vitro.
Preferences for phosphorylation sites in the retinoblastoma protein of D-type cyclin-dependent kinases, Cdk4 and Cdk6, in vitro.
Journal of biochemistry (2005-04-06)
Tohru Takaki, Kazuhiro Fukasawa, Ikuko Suzuki-Takahashi, Kentaro Semba, Masatoshi Kitagawa, Yoichi Taya, Hiroshi Hirai
PMID15809340
RÉSUMÉ
D-type cyclin-dependent kinases (Cdk4 and Cdk6) regulate the G1 to S phase progression of the mammalian cell cycle. It has been suggested that Cdk4 and Cdk6 may have distinct functions in vivo, even though they are indistinguishable biochemically. Here we show that although these Cdks phosphorylate multiple residues in pRB, they do so with different residue selectivities in vitro; Thr821 and Thr826 are preferentially phosphorylated by Cdk6 and Cdk4, respectively. This raises the possibility different substrate specificities lead to their different roles in the regulation of cellular events. Furthermore, our results indicate the new concept that Cdk itself contributes to substrate recognition.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CDK6/CyclinD1, active, His/GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution