- Stereoselective synthesis of 2-acetamido-1,2-dideoxyallonojirimycin (DAJNAc), a new potent hexosaminidase inhibitor.
Stereoselective synthesis of 2-acetamido-1,2-dideoxyallonojirimycin (DAJNAc), a new potent hexosaminidase inhibitor.
Organic letters (2013-06-27)
Alex de la Fuente, Ruben Martin, Teresa Mena-Barragán, Xavier Verdaguer, José M García Fernández, Carmen Ortiz Mellet, Antoni Riera
PMID23802126
RÉSUMÉ
A practical synthesis of the previously unreported N-acetyl-D-allosamine glycomimetic DAJNAc is described. The reaction sequence involves Pd-catalyzed allylic substitution by phthalimide in an azaheterobicyclic scaffold as the key step. The new iminosugar resulted in being a stronger β-N-acetylglucosaminidase (human placenta) competitive inhibitor than the D-gluco (DNJNAc) and D-galacto (DGJNAc) stereoisomers.
MATÉRIAUX