Merck
  • Simultaneous determination of glucocorticoids in plasma or urine by high-performance liquid chromatography with precolumn fluorimetric derivatization by 9-anthroyl nitrile.
Simultaneous determination of glucocorticoids in plasma or urine by high-performance liquid chromatography with precolumn fluorimetric derivatization by 9-anthroyl nitrile.
Journal of chromatography. B, Biomedical sciences and applications (1998-04-29)
N Shibata, T Hayakawa, K Takada, N Hoshino, T Minouchi, A Yamaji
RÉSUMÉ
A new method for simultaneous determination of glucocorticoids (GCs) in plasma or urine by high-performance liquid chromatography (HPLC) with fluorimetric detection has been developed. Following extraction with ethyl acetate using a reversed-phase disposable cartridge, the six GCs [cortisol (F), cortisone (E), prednisolone (PL), prednisone (PN), 6beta-hydroxycortisol (6beta-OHF) and 6beta-hydroxyprednisolone (6beta-OHP)] and an internal standard (6beta-hydroxycotortisone) were derivatized by treatment with 9-anthroyl nitrile (9-AN) in a mixture of basic catalysts (triethylamine and quinuclidine) to give the fluorescent esters through the 21-hydroxyl group. The GC derivatives so obtained were then cleaned by a straight-phase disposable cartridge and chromatographed on a straight-phase column with an isocratic HPLC technique. The fluorescence derivatives of the GCs, including the internal standard, were separated as clear single peaks and no interfering peaks were observed on the chromatograms. The lower limits of detection for F, E, PL and PN in plasma or urine were 0.1 ng/ml and those for 6beta-OHF and 6beta-OHP in plasma or urine were 0.5 ng/ml, at a signal-to-noise ratio of 3. The analytical recovery of known amounts of the GCs added to plasma or urine were almost 100%. This method can be applied to the determination of plasma or urinary F in renal transplant patients who received PL and can be applied for other metabolic investigations in relation to the change in blood pressure via 11beta-hydroxysteroid dehydrogenase or in hepatic metabolizing via CYP3A4.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
9-Anthracenecarbonitrile, 97%