Merck
  • 7H-Dibenzo[c,g]carbazole and 5,9-dimethyldibenzo[c,g]carbazole exert multiple toxic events contributing to tumor promotion in rat liver epithelial 'stem-like' cells.
7H-Dibenzo[c,g]carbazole and 5,9-dimethyldibenzo[c,g]carbazole exert multiple toxic events contributing to tumor promotion in rat liver epithelial 'stem-like' cells.
Mutation research (2006-01-13)
Jan Vondrácek, Lenka Svihálková-Sindlerová, Katerina Pencíková, Pavel Krcmár, Zdenek Andrysík, Katerina Chramostová, Sona Marvanová, Zuzana Valovicová, Alois Kozubík, Alena Gábelová, Miroslav Machala
RÉSUMÉ
Immature liver progenitor cells have been suggested to be an important target of hepatotoxins and hepatocarcinogens. The goal of the present study was to assess the impact of 7H-dibenzo[c,g]carbazole (DBC) and its tissue-specific carcinogenic N-methyl (N-MeDBC) and 5,9-dimethyl (DiMeDBC) derivatives on rat liver epithelial WB-F344 cells, in vitro model of liver progenitor cells. We investigated the cellular events associated with both tumor initiation and promotion, such as activation of aryl hydrocarbon receptor (AhR), changes in expression of enzymes involved in metabolic activation of DBC and its derivatives, effects on cell cycle, cell proliferation/apoptosis and inhibition of gap junctional intercellular communication (GJIC). N-MeDBC, a tissue-specific sarcomagen, was only a weak inhibitor of GJIC or inducer of AhR-mediated activity, and it did not affect either cell proliferation or apoptosis. DBC was efficient GJIC inhibitor, while DiMeDBC manifested the strongest AhR inducing activity. Accordingly, DiMeDBC was also the most potent inducer of cytochrome P450 1A1 (CYP1A1) and CYP1A2 expression among the three compounds tested. Both DBC and DiMeDBC induced expression of CYP1B1 and aldo-keto reductase 1C9 (AKR1C9). N-MeDBC failed to significantly upregulate CYP1A1/2 and it only moderately increased CYP1B1 or AKR1C9. Only the potent liver carcinogens, DBC and DiMeDBC, caused a significant increase of p53 phosphorylation at Ser15, an increased accumulation of cells in S-phase and apoptosis at micromolar concentrations. In addition, DiMeDBC was found to stimulate cell proliferation of contact-inhibited WB-F344 cells at 1 microM concentration, which is a mode of action that might further contribute to its hepatocarcinogenicity. The present data seem to suggest that the AhR activation, induction of enzymes involved in metabolic activation, inhibition of GJIC or stimulation of cell proliferation might all contribute to the hepatocarcinogenic effects of DBC and DiMeDBC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
7H-Dibenzo[c,g]carbazole, BCR®, certified reference material