Merck
Tandem electrophilic cyclization-[3+2] cycloaddition-rearrangement reactions of 2-alkynylbenzaldoxime, DMAD, and Br2.
The Journal of organic chemistry (2008-12-06)
Qiuping Ding, Zhiyong Wang, Jie Wu
RÉSUMÉ
Tandem electrophilic cyclization-[3+2] cycloaddition-rearrangement reactions of 2-alkynylbenzaldoximes, DMAD, and bromine are described, which afford the unexpected isoquinoline-based azomethine ylides in good to excellent yields. The products could be further elaborated via palladium-catalyzed cross-coupling reactions to generate highly functionalized isoquinoline-based stable azomethine ylides.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%