Merck
Construction and characterization of a fusion β-1,3-1,4-glucanase to improve hydrolytic activity and thermostability.
Biotechnology letters (2011-07-08)
Juntao Sun, Hongxin Wang, Wenping Lv, Chaoyang Ma, Zaixiang Lou, Yixing Dai
RÉSUMÉ
A new fusion gene (Bgl-licMB), encoding β-1,3-1,4-glucanase both from Bacillus amyloliquefaciens (Bgl) and Clostridium thermocellum (licMB), was constructed via end-to-end fusion and expressed in Escherichia coli to improve hydrolytic activity and thermostability of β-1,3-1,4-glucanase. The results of enzymatic properties showed that the catalytic efficiency (K(cat)/K(m)) of the fusion enzyme for oat β-glucan was 2.7 and 20-fold higher than that of the parental Bgl and licMB, respectively, and that the fusion enzyme can retain more than 50% of activity following incubation at 80°C for 30 min, whereas the residual activities of Bgl and licMB were both less than 30%. These properties make this particular β-1,3-1,4-glucanase a good candidate for application in brewing and animal-feed industries.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-Glucanase from Trichoderma longibrachiatum
Sigma-Aldrich
β-Glucanase from Aspergillus niger, powder, dark brown, ~1 U/mg