Merck
  • Organocatalytic ring expansion of beta-lactams to gamma-lactams through a novel N1-C4 bond cleavage. direct synthesis of enantiopure succinimide derivatives.
Organocatalytic ring expansion of beta-lactams to gamma-lactams through a novel N1-C4 bond cleavage. direct synthesis of enantiopure succinimide derivatives.
Organic letters (2005-08-27)
Benito Alcaide, Pedro Almendros, Gema Cabrero, M Pilar Ruiz
RÉSUMÉ
Tetrabutylammonium cyanide (20 mol %) catalyzes ring expansion of 4-(arylimino)methylazetidin-2-ones 2 to 5-aryliminopyrrolidin-2-ones 3 through a novel N1-C4 bond cleavage of the beta-lactam nucleus. New, efficient one-pot protocols to enantiopure succinimide derivatives 3 and 4 from beta-lactam aldehydes 1 have also been developed. [reaction: see text]
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%