Merck
Synthesis of α-Aryl Secondary Amides via Nickel-Catalyzed Reductive Coupling of Redox-Active Esters.
Organic letters (2022-04-27)
Alexis L Gabbey, Nicholas W M Michel, Jonathan M E Hughes, Louis-Charles Campeau, Sophie A L Rousseaux
RÉSUMÉ
The transition-metal-catalyzed α-arylation of secondary amides remains a synthetic challenge due to the presence of a free N-H bond. We report a strategy to synthesize secondary α-aryl amides via a Ni-catalyzed reductive arylation of redox-active N-hydroxyphthalimide (NHP) esters of malonic acid half amides. This transformation proceeds under mild conditions and displays excellent chemoselectivity for amide α-arylation in the presence of other enolizable carbonyls. The NHP ester substrates are readily prepared from Meldrum's acid.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ni(bpy)Cl2