Merck
Functional Dissection of the Enhancer Repertoire in Human Embryonic Stem Cells.
Cell stem cell (2018-07-24)
Tahsin Stefan Barakat, Florian Halbritter, Man Zhang, André F Rendeiro, Elena Perenthaler, Christoph Bock, Ian Chambers
RÉSUMÉ
Enhancers are genetic elements that regulate spatiotemporal gene expression. Enhancer function requires transcription factor (TF) binding and correlates with histone modifications. However, the extent to which TF binding and histone modifications functionally define active enhancers remains unclear. Here, we combine chromatin immunoprecipitation with a massively parallel reporter assay (ChIP-STARR-seq) to identify functional enhancers in human embryonic stem cells (ESCs) genome-wide in a quantitative unbiased manner. Although active enhancers associate with TFs, only a minority of regions marked by NANOG, OCT4, H3K27ac, and H3K4me1 function as enhancers, with activity markedly changing under naive versus primed culture conditions. We identify an enhancer set associated with functions extending to non-ESC-specific processes. Moreover, although transposable elements associate with putative enhancers, only some exhibit activity. Similarly, within super-enhancers, large tracts are non-functional, with activity restricted to small sub-domains. This catalog of validated enhancers provides a valuable resource for further functional dissection of the regulatory genome.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Roche
Cocktail d′inhibiteurs de protéases cOmplete, Tablets provided in EASYpacks
Sigma-Aldrich
Facteur inhibiteur de la leucémie, humain, 10 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture