Merck
Influence of 4-guanidinobutyric acid as coadsorbent in reducing recombination in dye-sensitized solar cells.
The journal of physical chemistry. B (2006-07-21)
Zhipan Zhang, Shaik M Zakeeruddin, Brian C O'Regan, Robin Humphry-Baker, Michael Grätzel
RÉSUMÉ
Dye-sensitized solar cells based on nanocrystalline TiO(2) have been fabricated with an amphiphilic ruthenium sensitizer [Ru (4,4'-dicarboxylic acid-2,2'-bipyridine) (4,4'-bis(p-hexyloxystyryl)-2,2'-bipyridine)(NCS)(2)], coded as K-19, and 4-guanidinobutyric acid (GBA) as coadsorbent. The cells showed a approximately 50 mV increase in open-circuit voltage and a similar current in comparison with cells without GBA cografting. The performance of both types of devices was evaluated on the basis of their photocurrent-voltage characteristics, dark current measurements, cyclic voltammetry, electrochemical impedance spectroscopy, and phototransient decay methods. The results indicate that GBA shifted the conduction band of TiO(2) toward a more negative potential and reduced the interfacial charge-transfer reaction from conduction band electrons to triiodide in the electrolyte (also known as the back reaction). In addition, the devices with GBA cografting showed an excellent stability with a power conversion efficiency of approximately 8% under simulated full sunlight (air mass 1.5, 100 mW cm(-2)) during visible light soaking at 60 degrees C.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Guanidinobutyric acid, ≥98%